missing translation for 'onlineSavingsMsg'
Learn More

Insulin Antibody (rIRDN/8546), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20705UV

619.65 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18564349

  • 728.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insulin
Monoclonal
DyLight 350
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Recombinant full-length human Insulin protein
0.1 mL
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
3630
Store at 4°C in the dark.
IgG2a κ
Immunohistochemistry (Paraffin)
rIRDN/8546
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.