missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Integrin alpha 5 beta 1 Antibody (volociximab) - Chimeric, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | Integrin alpha 5 beta 1 |
|---|---|
| Clone | volociximab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046635
|
Novus Biologicals™
NBP3-28753-100ug |
100 μg |
326.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045197
|
Novus Biologicals™
NBP3-28753-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046386
|
Novus Biologicals™
NBP3-28753-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Integrin alpha 5 beta 1 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Angiogenesis, Cancer, Immunology | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| 3678 | |
| IgG4 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| volociximab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A | |
| Integrin a5b1 (ITGA5 & ITGB1) | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title