Learn More
Integrin alpha-5 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10569-1-AP-20UL
 View more versions of this product
Product Code. 16877033
  • 145.00€ / 20µL
Description
Integrin alpha-5, also known as FNRA or VLA5A, is a protein that in humans is encoded by the ITGA5 gene. The product of this gene belongs to the integrin alpha chain family. Integrins are integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signaling.
Specifications
Specifications
Integrin alpha-5
Polyclonal
Unconjugated
ITGA5
CD49e, FNRA, Integrin alpha 5, Integrin alpha F, ITGA5, VLA 5, VLA5A
Rabbit
Antigen Affinity Chromatography
RUO
16402, 3678
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08648, P11688
ITGA5
Integrin alpha-5 Fusion Protein Ag0860
20 μL
Primary
Non-human Primate, Human, Mouse