missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 6 beta 1 Antibody (2548B), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB7809MFV500
This item is not returnable.
View return policy
Description
Integrin alpha 6 beta 1 Monoclonal antibody specifically detects Integrin alpha 6 beta 1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Integrin alpha 6 beta 1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Flow Cytometry, CyTOF | |
| 2548B | |
| Flow Cytometry, CyTOF-ready | |
| CD49f, ITGA6B, VLA-6 | |
| Chinese Hamster Ovary cell line, CHO-derived Human Integrin alpha 6 beta 1 heterodimer, Phe24-Ser1012(Integrin alpha 6) and Gln21-Asp728 (Integrin beta1), Accession # NP_000201 | |
| RUO | |
| 3655 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction