missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (CL6972) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76524JF549
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| This antibody was developed using a recombinant protein derived from P16144, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3691 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL6972 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction