Learn More
Diversey™ IntelliCare Dispenser Hybrid
Product Code. 17673105
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17673105 White Each
17663105 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17673105 Supplier Diversey™ Supplier No. D7524180
  • ONLINE EXCLUSIVE PRICE
    148.50€
    132.64€ / Pack of 1
    Save 15.86€
    10% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The IntelliCare Hybrid dispensing system can address all hand care needs with a versatile, modern, and durable platform that is compatible with both liquid/gels and foam products.
  • Dispenses 1300 mL bottles of hand soap and 1200 mL of alcohol based hand sanitizer
  • Can dispense 0.4 mL or 1.00 mL per push
  • Hybrid dispenser provides an automatic switch from touchless to manual if batteries died before replacement
  • Near empty product alarm and low battery signal
  • Locking dispenser contains a translucent window for product visibility without product tampering
Specifications
Product Type Hybrid Dispenser (Manual/Automatic)
Color White