Description
- The Interface I-300 is the central control unit for all process parameters of a Rotavapor R-300 system.
- This includes rotation speed, temperature (heating, cooling, vapor) and pressure.
- Various distinct operating modes (e.g. automatic, dynamic, manual distillation, drying etc.) increase efficiency and convenience of rotary evaporation processes.
Specifications
Specifications
| Material | Plastic |
| Type | User Interface |