Learn More
BUCHI Interface I-300 Pro
Product Code. 15500600 Shop All Buchi Products
Change view
Click to view available options
Type: User Interface
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15500600 Type: User Interface
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15500600 Supplier BUCHI Supplier No. 11I300P10
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The Interface I-300 Pro controls all distillation parameters of a Rotavapor R-300 system with various operating modes, touch screen control, recording and charting.
  • The Interface I-300 is the central control unit for all process parameters of a Rotavapor R-300 system.
  • This includes rotation speed, temperature (heating, cooling, vapor) and pressure.
  • Various distinct operating modes (e.g. automatic, dynamic, manual distillation, drying etc.) increase efficiency and convenience of rotary evaporation processes.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Type User Interface