missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interferon alpha Mouse anti-Human, Clone: 4A2A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66162-1-IG-20UL
This item is not returnable.
View return policy
Description
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.Specifications
| Interferon alpha | |
| Monoclonal | |
| 1.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01562 | |
| IFNA1 | |
| Interferon alpha Fusion Protein Ag12533 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 4A2A11 | |
| Unconjugated | |
| IFNA1 | |
| IFL, IFN, IFN ALPHA, IFN alpha 1/13, IFNA1, IFNA13, Interferon alpha 1/13, Interferon alpha D, interferon, alpha 1, Interferon-alpha1, LeIF D | |
| Mouse | |
| Protein A | |
| RUO | |
| 3439 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction