Learn More
Tuttnauer Internal Printer for D Line Autoclaves
Product Code. 17889538 Shop All Tuttnauer Products
Change view
Click to view available options
For Use With (Equipment):
D Line Autoclaves
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17889538 D Line Autoclaves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17889538 Supplier Tuttnauer Supplier No. PPR0002
  • ONLINE EXCLUSIVE PRICE
    1202.25€
    1046.69€ / Pack of 1
    Save 155.56€
    12% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
missing translation for 'validMainframeMsg'
Internal Printer for all D line units.
  • Internal Printer for cycle documentation.
  • For electronic models only.
  • DPU20 available for 2540 models only.
  • DPU30 available for all other models.
Specifications
For Use With (Equipment) D Line Autoclaves
Product Type Printer