Learn More
Brand™ Blaubrand™ IntraEND™ Disposable Micropipets
Product Code. 15252599 Shop All Brand Products
Change view
Click to view available options
:
Capacity: 1μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15252599 Capacity: 1μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15252599 Supplier Brand™ Supplier No. 709101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Comprised of disposable glass without ring mark. BRAND™ Blaubrand™ IntraEND™ Disposable Micropipets are in compliance with DIN ISO 7550 and CE-marked in accordance with IVD-Directive 98/79. Volume is defined by end-to-end filling. Volumes greater than 5μL include DE-M marking.
  • Comprised of disposable glass without ring mark.
  • Complies with DIN ISO 7550.
  • CE Marked in accordance with IVD-Directive 98/79
  • Calibrated to contain (TC, In.)
  • Volume is defined by end-to-end filling
  • Volumes greater than 5μL include DE-M marking.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ≤±0.5%
Calibration To Contain
Certifications/Compliance DIN ISO 7550, IVD-Directive 98/79 CE
Material Glass
Quantity 250/Box
Volume (Metric) 1 μL
Length (Metric) 32 mm
Disposable Single-use