Learn More
INTS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16130-1-AP-20UL
 View more versions of this product
Product Code. 16875644
  • 145.00€ / 20µL
Description
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.
Specifications
Specifications
INTS4
Polyclonal
Unconjugated
INTS4
INT4, integrator complex subunit 4, INTS4, MST093
Rabbit
Antigen Affinity Chromatography
RUO
101861, 308837, 92105
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CIM8, Q96HW7
Ints4
INTS4 Fusion Protein Ag9116
20 μL
Primary
Mouse, Rat, Human