Learn More
Invitrogen™ EveryPrep™ Universal Vacuum Manifold
Product Code. 10777985
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10777985 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10777985 Supplier Invitrogen™ Supplier No. K211101
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids.
The EveryPrep™ Universal Vacuum Manifold is designed for rapid manual purification of nucleic acids. It offers the ability to directly elute from the manifold using the new ChargeSwitch™ Pro Filter Plasmid Mini, Midi and Maxi Kits and also many other competitors' vacuum assisted spin column preps and filter plates. The EveryPrep™ Universal Vacuum Manifold replaces and consolidates all previous manifolds into one space saving benchtop unit. The manifold reduces sample handling to a minimum by allowing direct parallel and simultaneous processing of up to 96 samples using collection plates, deep-well blocks, or microtubes. The use of a vacuum eliminates the need for time-consuming centrifugation. The manifold will consist of a dual chamber construct fashioned out of aluminum with interchangeable top plates of a luer-lock tapped top plate, a 96-well top plate, a mini column elution plate and a maxi column elution plate.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuum Manifold
Content And Storage 1 vacuum manifold. Store at room temperature.
Quantity 1 each
Shipping Condition Room Temperature