Learn More
Invitrogen™ PureLink™ 96 Receiver Plates
Description
Includes
The PureLink 96 Plasmid Purification System is available as a complete kit and includes filter plates, receiver plates, air porous tape, foil tape, growth blocks, lysis buffer, TE buffer, cell suspension buffer, and RNase A.
- Centrifugation method eliminates cross-contamination
- PureLink 96 Plasmid Purification System is designed for high-throughput isolation of high-copy number plasmids
- PureLink 96 Plasmid Purification Systems protocol, up to 96 plasmid DNA samples can be simultaneously purified
- Novel technology eliminates time-consuming cell harvesting and alkaline lysis steps, saving time and effort
- Unique centrifugation-based format of the PureLink 96 Plasmid Purification System eliminates opportunities for cross-contamination between wells, critical factor in PCR and microarrays
- Isolated plasmid DNA is ideal for use in automated sequencing reactions, offering superior read lengths and data quality
- Samples may be processed 96 at time or in smaller increments
- PureLink 96 Plasmid Purification System is compatible with variety of centrifuges, robots, pipetting devices, automated sequencers, and sequencing chemistries
- PureLink 384 Plasmid Purification Plate is designed for ultra high throughput DNA isolation
- By utilizing same technology as PureLink 96 Plasmid Purification System, the PureLink 384 Plasmid Purification Plate reduces time and effort while reliably isolating high-quality plasmid DNA for DNA sequencing, PCR, and restriction digestion
- PureLink 96: Sample size 1.3 mL culture; Typical yields up to 3–5 μg
- PureLink 384: Sample size 200 μL culture; Typical yields up to 2–3 μg
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | The PureLink™ 96 Plasmid Purification System is available as a complete kit and includes filter plates, receiver plates, air porous tape, foil tape, growth blocks, lysis buffer, TE buffer, cell suspension buffer, and RNase A. Components are also available separately. PureLink™ 384 Plasmid Purification Plates are available in packs of 25. PureLink™ 384 Pre-Lysis Buffer is also available separately. Store at room temperature. All components are guaranteed stable for 2 years when properly stored. |
| Format | 96-well Plate |
| For Use With (Equipment) | Robotic Liquid Handling Systems |
| For Use With (Application) | Automated Sequencing Reaction |
| Plate Type | Receiver Plate |
| Product Line | PureLink |
| Product Type | Receiver Plate |
| Quantity | 50 Plates |
| Shipping Condition | Room Temperature |