Learn More
Invitrogen™ UltraPure™ Ethidium Bromide, 0.625 mg/mL
Product Code. 15361527
Change view
Click to view available options
Detection Method:
Gel Electrophoresis
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detection Method unitSize
15361527 Gel Electrophoresis Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15361527 Supplier Invitrogen™ Supplier No. A25645
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Fluorescent dye used to detect nucleic acids in agarose gels and cesium chloride (CsCl) gradient
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 x 1 mL ethidium bromide dropper (0.625 mg/mL)
Store at room temperature.
Detection Location In-Gel Detection
Detection Method Gel Electrophoresis
Label or Dye Ethidium Bromide
Shipping Condition Room Temperature
Target Molecule DNA & RNA
Product Line UltraPure
Product Type Ethidium Bromide