missing translation for 'onlineSavingsMsg'
Learn More

Iodine (Iodine-Iodide), 0.00500 N (N/200), Ricca Chemical

Product Code. 31750933
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750933 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750933 Supplier Ricca Chemical Company Supplier No. R39705004C
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7732-18-5, 7681-11-0, 7553-56-2
Physical Form Liquid
Quantity 4 L
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
Solubility Information Solubility in Water: Infinite
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Grade Laboratory
Odor Characteristic weak Iodine odor
Packaging Amber Glass Bottle
Appearance Dark brown liquid
Chemical Name or Material Iodine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.