missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodobenzene-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Iodobenzene-d5 |
| CAS | 7379-67-1 |
| Molecular Formula | C6 2H5 I |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6-iodo-, Benzene-d5, iodo- (7 CI,8 CI,9 CI), 6-Iodobenzene-1,2,3,4,5-d5, Iodobenzene-d5, Iodobenzene D5 |
| InChI Formula | InChI=1 S/C6H5I/c7-6-4-2-1-3-5-6/h1-5 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(I)c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-iodobenzene |
| Molecular Weight (g/mol) | 209.04 |
| Formula Weight | 208.975 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction