missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodomesitylene, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Iodomesitylene |
| CAS | 4028-63-1 |
| Molecular Formula | C9H11I |
| Quantity | 100 mg |
| Synonym | Mesityl Iodide, NSC 68044, 2-Iodo-1,3,5-trimethylbenzene, 1,3,5-Trimethyl-2-iodobenzene, 1-Iodo-2,4,6-trimethylbenzene, 2,4,6-Trimethyliodobenzene, 2,4,6-Trimethylphenyl Iodide, 2-Iodomesitylene, Iodo-2,4,6-trimethylbenzene |
| InChI Formula | InChI=1S/C9H11I/c1-6-4-7(2)9(10)8(3)5-6/h4-5H,1-3H3 |
| SMILES | Cc1cc(C)c(I)c(C)c1 |
| IUPAC Name | 2-iodo-1,3,5-trimethylbenzene |
| Molecular Weight (g/mol) | 246.09 |
| Formula Weight | 245.9905 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction