Learn More
Mitutoyo™ ABSOLUTE Digimatic Caliper
Product Code. 15666424
Encompass_Preferred
Change view
Click to view available options
Range:
0 to 200 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
15666424 0 to 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15666424 Supplier Mitutoyo™ Supplier No. 50070720
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
  • Designed for use in challenging work environments.
  • Extremely resistant to coolants and lubricants.
  • Available with or without data output
Specifications
Display Digital
Includes Without thumb roller
IP Rating IP66
Range 0 to 200 mm