Learn More
Grant Instruments™ IQ/OQ Documentation
Product Code. 16468300
Change view
Click to view available options
For Use With (Equipment):
UVC/T-M-AR General Purpose PCR UV Cabinet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16468300 UVC/T-M-AR General Purpose PCR UV Cabinet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16468300 Supplier Grant Instruments™ Supplier No. IQOQ UVC/TMAR
  • ONLINE EXCLUSIVE PRICE
    461.00€
    430.20€ / Pack of 1
    Save 30.80€
    6% Off
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
  • Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
  • Operational Qualification (OQ): instrument is functioning in accordance with its specification
  • Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) UVC/T-M-AR General Purpose PCR UV Cabinet
Product Type IQ/OQ Documentation