missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Antibody (2H9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37575-0.025ml
This item is not returnable.
View return policy
Description
IRAK4 Monoclonal specifically detects IRAK4 in Human, Mouse, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| IRAK4 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| IRAK4 | |
| Purified recombinant fragment of human IRAK4 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Monkey | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 2H9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
| Mouse | |
| 52 kDa | |
| 0.025 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 51135 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction