missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99534-100ul
This item is not returnable.
View return policy
Description
IRAK4 Polyclonal antibody specifically detects IRAK4 in Human samples. It is validated for Western Blot, ELISA
Specifications
| IRAK4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
| Produced in rabbits immunized with purified, recombinant Human IRAK4 (Uniprot#: Q9NWZ3-1; Met1-Ser460) | |
| 100 μg | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 51135 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction