missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRE1 alpha Antibody (142), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90017F
This item is not returnable.
View return policy
Description
IRE1 alpha Monoclonal antibody specifically detects IRE1 alpha in Human samples. It is validated for ELISA
Specifications
| IRE1 alpha | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 142 | |
| ELISA | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IRE1 alpha (O75460-1; Pro465-Leu977). | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Diabetes Research, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction, Unfolded Protein Response | |
| 2081 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction