missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Antibody (MUM1/8274R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20718IR
This item is not returnable.
View return policy
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IRF4 | |
| Monoclonal | |
| DyLight 755 | |
| interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
| Recombinant fragment (corresponding to C-terminal) of IRF4 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 3662 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MUM1/8274R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion