Learn More
IRF5 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507252S
Description
IRF5 Monoclonal antibody specifically detects IRF5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IRF5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13568 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3663 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G7 | |
| Unconjugated | |
| IRF5 | |
| AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
| Full length human recombinant protein of human IRF5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.