Learn More
IRF5 Mouse anti-Human, Unconjugated, Clone: 3A4A10, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | IRF5 |
|---|---|
| Clone | 3A4A10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889075
|
Proteintech
66835-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879075
|
Proteintech
66835-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IRF5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IRF5. This antibody recognizes human, mouse antigen. The IRF5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IRF5 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 3663 | |
| Primary |
| 3A4A10 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.