missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF8 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1093
This item is not returnable.
View return policy
Description
IRF8 Polyclonal specifically detects IRF8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| IRF8 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ICSBPH-ICSBP, interferon consensus sequence binding protein 1, Interferon consensus sequence-binding protein, interferon regulatory factor 8, IRF-8ICSBP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3394 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:8000 | |
| NP_002154 | |
| IRF8 | |
| Peptide with sequence C-QRSFFRENQQITV corresponding to C-Terminus according to NP_002154.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Cow, and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction