missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Acetylacetonate, TRC

Product Code. 30408644
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408644 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408644 Supplier Toronto Research Chemicals Supplier No. TRCI7752232.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Iron(III) Acetylacetonate
CAS 14024-18-1
Molecular Formula C15H21FeO6
Quantity 2.5 g
Synonym (Acetylacetonato)iron(III), Acetope Fe(II), Ferric Acetylacetonate, Ferric tris(Acetoacetonate), Ferric tris(Acetylacetonate), Iron Acetylacetonate, Iron Triacetylacetonate, Iron Tris(2,4-pentanedionate), Iron tris(Acetoacetonate), Iron tris(acetylacetonate), Iron(3+) Acetylacetonate, Iron(III) Acetylacetonate, Iron(III) tris(2,4-pentanedionate), NSC 43622, Nacem Ferric, Nacem Ferric Iron, Nacem Iron, Nasem Ferric Iron, Tris(2,4-pentanedionato)iron, Tris(acetylacetonato)iron, Tris(acetylacetone)iron
InChI Formula InChI=1S/3C5H8O2.Fe/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;
SMILES CC1=O[Fe+3]23(O=C(C)C=C(C)[O-]2)(O=C(C)C=C(C)[O-]3)[O-]C(=C1)C
Molecular Weight (g/mol) 353.17
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.