missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Acetylacetonate, TRC
Specifications
Specifications
| Chemical Name or Material | Iron(III) Acetylacetonate |
| CAS | 14024-18-1 |
| Molecular Formula | C15H21FeO6 |
| Quantity | 250 mg |
| Synonym | (Acetylacetonato)iron(III), Acetope Fe(II), Ferric Acetylacetonate, Ferric tris(Acetoacetonate), Ferric tris(Acetylacetonate), Iron Acetylacetonate, Iron Triacetylacetonate, Iron Tris(2,4-pentanedionate), Iron tris(Acetoacetonate), Iron tris(acetylacetonate), Iron(3+) Acetylacetonate, Iron(III) Acetylacetonate, Iron(III) tris(2,4-pentanedionate), NSC 43622, Nacem Ferric, Nacem Ferric Iron, Nacem Iron, Nasem Ferric Iron, Tris(2,4-pentanedionato)iron, Tris(acetylacetonato)iron, Tris(acetylacetone)iron |
| InChI Formula | InChI=1S/3C5H8O2.Fe/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-; |
| SMILES | CC1=O[Fe+3]23(O=C(C)C=C(C)[O-]2)(O=C(C)C=C(C)[O-]3)[O-]C(=C1)C |
| Molecular Weight (g/mol) | 353.17 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?