missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Iron(III) Chloride |
| CAS | 7705-08-0 |
| Molecular Formula | Cl3 Fe |
| Quantity | 25 g |
| Synonym | Bellair green plus, Clarfer, Clayfec, Danube clear classic, Donau klar classic, FC-Pix, Ferric chloride, Ferric trichloride, Ferrisol Cl 130, Flores martis, Iron chloride, Iron perchloride, Iron trichloride, NSC 135798, NSC 51150, Riedel 12322, Trifloc, ViscoStat Plus |
| InChI Formula | InChI=1S/3ClH.Fe/h3*1H;/q;;;+3/p-3 |
| SMILES | Cl[Fe](Cl)Cl |
| IUPAC Name | trichloroiron |
| Molecular Weight (g/mol) | 162.204 |
| Formula Weight | 160.8415 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?