missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Oxide, Solstice

Product Code. 15613160 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613160 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613160 Supplier Honeywell-Fluka Supplier No. 31005025G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powder, <5 μm,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1309-37-1
Physical Form Powder
Quantity 25 g
Molecular Formula Fe2O3
MDL Number MFCD00011008
UN Number NONH for all modes of transport
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES [Fe][Fe](=O)(=O)=O
IUPAC Name trioxodiiron
Molecular Weight (g/mol) 159.69
Formula Weight 159.69g/mol
Percent Purity ≥99%
Chemical Name or Material Iron(Iii) Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.