missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, Technical, Strip 200 X 15 X 0.5mm, Fisher Chemical

Product Code. 11488443 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 Pair
Packaging:
Plastic container
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11488443 1 Pair Plastic container 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11488443 Supplier Fisher Chemical Supplier No. I/1021/99
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7439-89-6
Melting Point 1535°C
Boiling Point 3000°C
Quantity 1 Pair
Packaging Plastic container
Vapor Pressure 1mmHg 1787
Form Solid
Molecular Formula Fe
MDL Number MFCD00010999
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES [Fe]
IUPAC Name iron
Molecular Weight (g/mol) 55.85
PubChem CID 23925
ChEBI CHEBI:82664
Formula Weight 55.84g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.