missing translation for 'onlineSavingsMsg'
Learn More

Iron, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15527994 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15527994 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15527994 Supplier Avantor J.T.Baker Supplier No. 222601

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7439-89-6
Quantity 500 g
Packaging Wide Mouth Amber Glass Bottle
Form Powder
Molecular Formula Fe
MDL Number MFCD00010999
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES [Fe]
IUPAC Name iron
Molecular Weight (g/mol) 55.85
PubChem CID 23925
ChEBI CHEBI:82664
Formula Weight 55.85
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Iron
Show More Show Less
compliance-icons
Product Identifier
  • Iron
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Iron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.