Learn More
ISA/Ca Model Ion Strength Adjustment Solutions, WTW™
Product Code. 11715214
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11715214 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11715214 Supplier WTW Supplier No. 140120
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.
TRUSTED_SUSTAINABILITY
Specifications
Packaging 1 Bottle with 250mL
Density 1.18g/cm3
Physical Form Liquid
Concentration 4 mol/L
Boiling Point 100°C
Solubility Miscible with water
Quantity 250 mL