missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ISL1 Recombinant Rabbit Monoclonal Antibody (1A1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538401
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.
Specifications
| ISL1 | |
| Recombinant Monoclonal | |
| 0.35 mg/mL | |
| PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 | |
| P61371 | |
| ISL1 | |
| A synthesized peptide derived from human Islet1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1A1 | |
| Unconjugated | |
| ISL1 | |
| Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3670 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering