missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Islet-1 Antibody (CL11651) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15121AF532
This item is not returnable.
View return policy
Description
Islet-1 Monoclonal antibody specifically detects Islet-1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Islet-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1 | |
| This antibody was developed using a synthetic peptide derived from P61371, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Stem Cell Markers | |
| 3670 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunocytochemistry/Immunofluorescence | |
| CL11651 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction