missing translation for 'onlineSavingsMsg'
Learn More

Iso Cephalomannine, TRC

Product Code. 17940945
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17940945 5 mg 5mg
17973335 10 mg 10mg
17971415 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17940945 Supplier Toronto Research Chemicals Supplier No. TRCI7895005MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173101-54-7
Molecular Formula C45 H53 N O14
Molecular Weight (g/mol) 831.9
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-12-[[(2E)-2-methyl-1-oxo-2-buten-1-yl]oxy]-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-12-[[(2E)-2-methyl-1-oxo-2-butenyl]oxy]-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)- (9CI), Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, 6,12b-bis(acetyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-12-[(2-methyl-1-oxo-2-butenyl)oxy]-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α(E),12aα,12bα]]-, 2-Debenzoyl-2-tigloylpaclitaxel, Isocephalomannine, Paclitaxel Imp A (EP)
SMILES C\C=C(/C)\C(=O)O[C@H]1[C@@H]2[C@@]3(CO[C@@H]3C[C@H](O)[C@@]2(C)C(=O)[C@H](OC(=O)C)C4=C(C)[C@H](C[C@]1(O)C4(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c5ccccc5)c6ccccc6)OC(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C45H53NO14/c1-9-23(2)40(53)59-38-36-43(8,30(49)20-31-44(36,22-56-31)60-26(5)48)37(51)35(57-25(4)47)32-24(3)29(21-45(38,55)42(32,6)7)58-41(54)34(50)33(27-16-12-10-13-17-27)46-39(52)28-18-14-11-15-19-28/h9-19,29-31,33-36,38,49-50,55H,20-22H2,1-8H3,(H,46,52)/b23-9+/t29-,30-,31+,33-,34+,35+,36-,38-,43+,44-,45+/m0/s1
Formula Weight 831.3466
Chemical Name or Material Iso Cephalomannine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.