missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Propyl-1,1,1,3,3,3-d6 Alcohol, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Propanol-1,1,1,3,3,3-D6 |
| CAS | 3976-29-2 |
| Molecular Formula | C3 D6 H2 O |
| Quantity | 5 g |
| Synonym | 2-Propan-1,1,1,3,3,3-d6-ol (9 CI), Isopropyl-2,2,2,2',2',2'-d6 alcohol (7 CI, 8 CI), 1,1,1,3,3,3-Hexadeutero-2-propanol, 2-Propanol-1,1,1,3,3,3-D6, iso-Propyl-1,1,1,3,3,3-d6 Alcohol |
| InChI Formula | InChI=1 S/C3H8O/c1-3(2)4/h3-4 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,3,3,3-hexadeuteriopropan-2-ol |
| Molecular Weight (g/mol) | 66.132 |
| Formula Weight | 66.0952 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction