Learn More
Brady™ ISO Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17861765 Shop All Brady Products
Change view
Click to view available options
Material:
PVC
Shape:
L-shaped
V-shaped
Style:
Class B
Class C
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Shape Style Unit Size
17861765 PVC L-shaped Class C Each
17831795 PVC V-shaped Class C Each
17821795 PVC L-shaped Class B Each
17841765 PVC V-shaped Class B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17861765 Supplier Brady™ Supplier No. 311226
  • ONLINE EXCLUSIVE PRICE
    29.00€
    26.16€ / Pack of 1
    Save 2.84€
    9% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Certifications/Compliance Class B
Alternate Part Number F/F018/NT/BIPVC-PHOLUMB-151X151-1
Finish Gloss
Material PVC
Quantity 1 Sign
Style Class C
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 5.94 in.
Width (English) 5.94 in.
Temperature Range (Metric) -40°C to +66°C
Shape L-shaped
Temperature Range (English) -40°F to +150°F
Languages Universal
Dimensions (L x W) 151 x 151 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color Photoluminescent on Red