Learn More
Brady™ ISO Safety Sign 'Wear Laboratory Coat,' English
Product Code. 17851715 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
50 x 150 mm
105 x 297 mm
150 x 450 mm
200 x 600 mm
210 x 148 mm
297 x 210 mm
420 x 297 mm
Material:
Polyester
Polypropylene
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Material Unit Size
17851715 105 x 297 mm Polypropylene Each
17881695 50 x 150 mm Polyester Each
17811705 105 x 297 mm Polyester Each
17831775 150 x 450 mm Polyester Each
17821705 200 x 600 mm Polyester Each
17801785 210 x 148 mm Polyester Each
17891775 297 x 210 mm Polyester Each
17831725 420 x 297 mm Polyester Each
17871785 50 x 150 mm Polypropylene Each
17871745 150 x 450 mm Polypropylene Each
17831755 200 x 600 mm Polypropylene Each
17861705 210 x 148 mm Polypropylene Each
17881785 297 x 210 mm Polypropylene Each
17881715 420 x 297 mm Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17851715 Supplier Brady™ Supplier No. 311381
  • ONLINE EXCLUSIVE PRICE
    12.40€
    11.17€ / Pack of 1
    Save 1.23€
    9% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Safety Sign to signify that a laboratory coat must be worn.
  • Provide visual warning that laboratory coat must be worn in the area
Specifications
Alternate Part Number M/M059/EN566/PP-297X105-1
Material Polypropylene
Quantity 1 Sign
Size (English) 4.13 x 11.69 in.
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 4.13 in.
Thickness 1.55 mm
Width (English) 11.69 in.
Temperature Range (Metric) -40°C to +120°C
Shape Rectangular
Temperature Range (English) -40°F to +248°F
Languages English
Dimensions (L x W) 105 x 297 mm
Displayed Text Wear Laboratory Coat
Product Type Safety Sign
Color White on Blue