missing translation for 'onlineSavingsMsg'
Learn More

Isobergapten, TRC

Product Code. 30138424
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138424 25 mg 25mg
30132904 2.5 mg 2.5mg
30004408 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138424 Supplier Toronto Research Chemicals Supplier No. TRCI77887525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 482-48-4
Molecular Formula C12 H8 O4
Molecular Weight (g/mol) 216.19
Synonym 5-Methoxyangelicin, Isobergaptene
IUPAC Name 5-methoxyfuro[2,3-h]chromen-2-one
SMILES COc1cc2occc2c3OC(=O)C=Cc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H8O4/c1-14-9-6-10-8(4-5-15-10)12-7(9)2-3-11(13)16-12/h2-6H,1H3
Formula Weight 216.0423
Chemical Name or Material Isobergapten
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.