missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isobutyltriphenylphosphonium bromide, 98+%, Thermo Scientific™

Product Code. 11453488
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453488 25 g 25g
11443488 5 g 5g
11463488 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453488 Supplier Alfa Aesar™ Supplier No. A17881.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22884-29-3
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00031593
InChI Key VIKPDPHNYHUZQW-UHFFFAOYSA-M
Synonym isobutyltriphenylphosphonium bromide, 2-methylpropyl triphenyl phosphanium bromide, phosphonium, 2-methylpropyl triphenyl-, bromide, 2-methylpropyl triphenylphosphanium bromide, acmc-1citv, isobutyltriphenylphosphoniumbromide, isobutyl triphenylphosphonium bromide, 2-methylpropyl triphenyl phosphonium bromide, isobutyl triphenyl phosphonium bromide, phosphonium, 2-methylpropyl triphenyl-,bromide 1:1
PubChem CID 2724571
SMILES [Br-].CC(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 198°C to 202°C
Quantity 25 g
Beilstein 4171228
IUPAC Name 2-methylpropyl(triphenyl)phosphanium;bromide
Formula Weight 399.32
Percent Purity ≥98%
Chemical Name or Material Isobutyltriphenylphosphonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.