Learn More
Isocitrate Dehydrogenase 2/IDH2 Antibody (S04-1B0), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19658-100ul
 View more versions of this product
Product Code. 18685583
  • 645.00€ / 100µL
Description
Isocitrate Dehydrogenase 2/IDH2 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 2/IDH2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Isocitrate Dehydrogenase 2/IDH2
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
A synthetic peptide of human Isocitrate Dehydrogenase 2/IDH2 (Uniprot # P48735)
100 μg
Primary
Human, Mouse, Rat
Purified
Western Blot
S04-1B0
Unconjugated
Rabbit
Affinity purified
RUO
3418
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.