missing translation for 'onlineSavingsMsg'
Learn More

Isohumulone, TRC

Product Code. 30006776
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006776 25 mg 25mg
30004134 5 mg 5mg
30005003 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006776 Supplier Toronto Research Chemicals Supplier No. TRCI79042025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25522-96-7
Molecular Formula C21 H30 O5
Molecular Weight (g/mol) 362.46
Synonym 3,4-Dihydroxy-5-(3-methyl-2-buten-1-yl)-2-(3-methyl-1-oxobutyl)-4-(4-methyl-1-oxo-3-penten-1-yl)-2-cyclopenten-1-one, Iso-n-humulone, IsoHopCO2N
IUPAC Name 3,4-dihydroxy-2-(3-methylbutanoyl)-5-(3-methylbut-2-enyl)-4-(4-methylpent-3-enoyl)cyclopent-2-en-1-one
SMILES CC(C)CC(=O)C1=C(O)C(O)(C(CC=C(C)C)C1=O)C(=O)CC=C(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H30O5/c1-12(2)7-9-15-19(24)18(16(22)11-14(5)6)20(25)21(15,26)17(23)10-8-13(3)4/h7-8,14-15,25-26H,9-11H2,1-6H3
Formula Weight 362.2093
Chemical Name or Material Isohumulone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.