Learn More
Labconco™ Isolation Valve
Product Code. 15183085 Shop All Labconco Corporation Products
Change view
Click to view available options
Type:
Isolation Valve
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15183085 Isolation Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15183085 Supplier Labconco™ Supplier No. 7761500
  • ONLINE EXCLUSIVE PRICE
    4210.00€
    3581.89€ / Pack of 1
    Save 628.11€
    14% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Shorten the time necessary to reestablish working vacuum levels and to provide a means for determining end point. Labconco™ Isolation Valve is for use with FreeZone™ Stoppering Tray Dryers and FreeZone™ Bulk Tray Dryers.
  • Isolates the Tray Dryer from the Freeze Dry System to shorten the time necessary to reestablish working vacuum levels and to provide a means for determining end point
  • Consists of a valve, valve handle, clamps, couplings and installation instructions
  • Mounts underneath the support stand included with the Tray Dryer
  • May be installed before or after the Tray Dryer is installed on the Freeze Dry System
  • For use with Stoppering Tray Dryers 7806020, 7806040 and 7806030 and Bulk Tray Dryers 7806060 and 7806070.
  • The Isolation Valve and 6-Port Tray Dryer Manifold 7726500 may not be field installed together.
Specifications
Type Isolation Valve
Height (English) 7.75 in.
Length (English) 8.65 in.
Shipping Weight (English) 5 lb.
Shipping Weight (Metric) 2.3 kg