missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinamide, 99%, ACROS Organics™

Product Code. 10446930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10446930 25g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10446930 Supplier Thermo Scientific Acros Supplier No. 122580250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1453-82-3
Melting Point 155°C to 158°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C6H6N2O
MDL Number MFCD00006432
Quantity 25 g
Beilstein 22, 46
Synonym isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q
InChI Key VFQXVTODMYMSMJ-UHFFFAOYSA-N
SMILES C1=CN=CC=C1C(=O)N
IUPAC Name pyridine-4-carboxamide
Molecular Weight (g/mol) 122.127
PubChem CID 15074
ChEBI CHEBI:6031
Formula Weight 122.13
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.