missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinamide, Mikromol™

Product Code. 17934166
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934166 100 mg 100mg
17912686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17934166 Supplier Mikromol Supplier No. MM0552.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Isonicotinamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isonicotinamide
CAS 1453-82-3
Analyte or Component Names Isonicotinamide
Shipping Condition Room Temperature
Molecular Formula C6 H6 N2 O
Certifications/Compliance ISO 17025
Synonym Isonicotinamide, 4-Pyridinecarboxamide, Isonicotinamide (6CI,7CI,8CI), 4-(Aminocarbonyl)pyridine, 4-Amidopyridine, 4-Carbamoylpyridine, 4-Pyridinecarboxylic acid amide, Isoniacinamide, Isonicotinic acid amide, NSC 82353, Pyridine-4-carbamide, gamma-Pyridinecarboxamide, Isoniazid Impurity B
InChI Formula InChI=1S/C6H6N2O/c7-6(9)5-1-3-8-4-2-5/h1-4H,(H2,7,9)
SMILES NC(=O)c1ccncc1
IUPAC Name pyridine-4-carboxamide
Molecular Weight (g/mol) 122.12
Quantity 100 mg
Formula Weight 122.048 g/mol
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.