Learn More
Isonicotinoyl chloride hydrochloride, 97%
Product Code. 11439852
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439852 250 g 250g
11419852 10 g 10g
11429852 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11439852 Supplier Thermo Scientific Alfa Aesar Supplier No. A10863.30
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39178-35-3
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00012830
InChI Key BNTRVUUJBGBGLZ-UHFFFAOYSA-N
Synonym isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
PubChem CID 12262826
IUPAC Name pyridine-4-carbonyl chloride;hydrochloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=NC=C1
Specifications
Melting Point 154°C to 162°C
Quantity 250 g
UN Number UN3261
Beilstein 3626052
Sensitivity Hygroscopic
Formula Weight 178.01
Percent Purity 98%
Chemical Name or Material Isonicotinoyl chloride hydrochloride