missing translation for 'onlineSavingsMsg'
Learn More

Isononylphenol, TRC

Product Code. 30138258
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138258 50 mg 50mg
17937149 10 mg 10mg
17918489 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138258 Supplier Toronto Research Chemicals Supplier No. TRCI42014050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11066-49-2
Molecular Formula C15 H24 O
Molecular Weight (g/mol) 220.35
Synonym 2-(7-Methyloctyl)phenol
IUPAC Name 2-(7-methyloctyl)phenol
SMILES CC(C)CCCCCCc1ccccc1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H24O/c1-13(2)9-5-3-4-6-10-14-11-7-8-12-15(14)16/h7-8,11-13,16H,3-6,9-10H2,1-2H3
Formula Weight 220.1827
Percent Purity >95%
Chemical Name or Material Isononylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.