missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl Acetate, TRC

Product Code. 30405413
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405413 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405413 Supplier Toronto Research Chemicals Supplier No. TRCI82278010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropenyl Acetate
CAS 108-22-5
Molecular Formula C5 H8 O2
Quantity 10 g
Synonym 1-Propen-2-ol, 2-acetate, 1-Propen-2-ol, acetate (6CI,8CI,9CI), 1-Acetoxy-1-methylethylene, 1-Methylvinyl acetate, 2-Acetoxy-1-propene, 2-Acetoxypropene, 2-Acetoxypropylene, Acetic acid isopropenyl ester, Isopropenyl acetate, NSC 2197, NSC 8262, Prop-1-en-2-yl acetate, Acetic acid-isopropenyl ester
InChI Formula InChI=1S/C5H8O2/c1-4(2)7-5(3)6/h1H2,2-3H3
SMILES CC(=C)OC(=O)C
IUPAC Name prop-1-en-2-yl acetate
Molecular Weight (g/mol) 100.1158
Formula Weight 100.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.