missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl bromoacetate, TRC

Product Code. 30405405
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405405 10 g 10g
30398109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405405 Supplier Toronto Research Chemicals Supplier No. TRCI87463510G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl bromoacetate
CAS 29921-57-1
Molecular Formula C5H9BrO2
Quantity 10 g
Synonym Propan-2-yl-2-bromoacetate, Acetic acid, 2-bromo-1-methylethyl ester
InChI Formula InChI=1S/C5H9BrO2/c1-4(2)8-5(7)3-6/h4H,3H2,1-2H3
SMILES CC(C)OC(=O)CBr
IUPAC Name propan-2-yl 2-bromoacetate
Molecular Weight (g/mol) 181.03
Formula Weight 179.9786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.